missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) sulfide, ultra dry, 99.98% (metals basis)

Manufacturer:  Alfa Aesar™ 1430009

 View more versions of this product

Catalog No. AA1430009



Description

Description

  • Ampouled under argon
Specifications

Specifications

Copper(II) sulfide
1317-40-4
CuS
14,2965
BWFPGXWASODCHM-UHFFFAOYSA-N
sulfanylidenecopper
14831
Powder
3.9g/mL to 4.6g/mL
Ampule
Air & Moisture Sensitive
10g
Ultra dry
(metals basis)
MFCD00016066
copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue
S=[Cu]
95.606
95.6
99.98%
Odorless
1.45
220°C (decomposition)

Chemical Identifiers

1317-40-4
95.606
BWFPGXWASODCHM-UHFFFAOYSA-N
14831
S=[Cu]
CuS
MFCD00016066
copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue
sulfanylidenecopper
Safety and Handling

Safety and Handling

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents