Please login to your online account to display your discounted pricing

Copper(II) Sulfate, Pentahydrate, ACS, 99.0 to 100.5%, EMD Millipore

$70.71 - $70.71

Chemical Identifiers

Chemical Name or Material Copper(II) Sulfate, Pentahydrate
Name Note EMSURE™, For Analysis
CAS 7758-99-8
Purity Grade ACS/EP/ISO/Reagent Grade
Physical Form Crystals
Assay Percent Range 99.0 to 100.5%
Molecular Formula CuO4S · 5H2O
Linear Formula CuSO4 · 5H2O
Formula Weight 249.68
Synonym Copper monosulfate pentahydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

M1027900250

 
emd millipore corporation
1027900250
250g Each for $70.71
Description & Specifications

Specifications

Formula Weight 249.68
Purity Grade ACS/EP/ISO/Reagent Grade
Physical Form Crystals
Color Blue
Quantity 250g
Density 2.284g/cm3 (20°C)
Synonym Copper monosulfate pentahydrate
Chemical Name or Material Copper(II) Sulfate, Pentahydrate
Linear Formula CuSO4 · 5H2O
Name Note EMSURE™, For Analysis
Assay Percent Range 99.0 to 100.5%
CAS 7758-99-8
Molecular Formula CuO4S · 5H2O
Recommended Storage Store without restrictions
Safety and Handling

Store without restrictions