Please login to your online account to display your discounted pricing

Copper(II) sulfate pentahydrate, 98+%, ACS reagent, ACROS Organics™


Quantity 5g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  423610050

 View more versions of this product

Catalog No. AC423610050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 249.68
Physical Form Fine Crystals
Quantity 5g
Color Blue
Assay 98.0 to 102.0%
Fieser 01,164; 02,89; 05,162; 06,141; 14,267
Insoluble Matter 0.005% max.
Linear Formula CuSO4·5H2O
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98+%
Molecular Formula CuO45H2O
Chemical Name or Material Copper(II) sulfate pentahydrate
CAS 7758-99-8
Packaging Plastic bottle
Iron (Fe) 30ppm max.
Nickel (Ni) 50ppm max.
Potassium (K) 100ppm max.
Calcium (Ca) 50ppm max.
Sodium (Na) 200ppm max.
Nitrogen Compounds 0.002% max.
Chlorine (Cl) 10ppm max.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 1 Warning

Specifications

Chemical Name or Material Copper(II) sulfate pentahydrate
CAS 7758-99-8
Purity Grade ACS Reagent Grade
Assay 98.0 to 102.0%
Assay Percent Range 98+%
Molecular Formula CuO45H2O
Linear Formula CuSO4·5H2O
Formula Weight 249.68
Accessories

Warning

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents