Learn More
Copper(II) sulfate pentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423615000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper(II) sulfate pentahydrate | |
7758-99-8 | |
Fine Crystals | |
98.0 to 102% (Iodometry) | |
CuSO4·5H2O | |
01,164; 02,89; 05,162; 06,141; 14,267 | |
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
copper;sulfate;pentahydrate | |
24463 | |
249.68 | |
ACS Reagent |
110.0°C | |
Blue | |
500 g | |
CuH10O9S | |
MFCD00149681 | |
15,2643 | |
Solubility in water: 320g/L (20°C). Other solubilities: soluble in methanol, slightly soluble in alcohol and glycerol, insoluble in ethanol | |
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O | |
249.68 | |
CHEBI:31440 | |
≥98% | |
Plastic bottle |
Chemical Identifiers
7758-99-8 | |
249.68 | |
JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
24463 | |
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
CuH10O9S | |
MFCD00149681 | |
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
CHEBI:31440 |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only