Learn More
Copper(II) sulfate, anhydrous, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 033308A3
Description
Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in the Fehling's solution and Benedict's solution to test for reducing sugars. Also, it is used to test protein in Biuret test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper(II) sulfate | |
7758-98-7 | |
Powder | |
CuO4S | |
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle | |
ARUVKPQLZAKDPS-UHFFFAOYSA-L | |
copper;sulfate | |
24462 | |
159.6 | |
Odorless | |
3.6 |
>560°C (decomposition) | |
Gray | |
2 kg | |
MFCD00010981 | |
Soluble in water and methanol. Insoluble in ethanol. | |
[O-]S(=O)(=O)[O-].[Cu+2] | |
159.602 | |
CHEBI:23414 | |
Reagent | |
Hygroscopic |
Chemical Identifiers
7758-98-7 | |
159.602 | |
ARUVKPQLZAKDPS-UHFFFAOYSA-L | |
24462 | |
copper;sulfate |
CuO4S | |
MFCD00010981 | |
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle | |
CHEBI:23414 | |
[O-]S(=O)(=O)[O-].[Cu+2] |
Safety and Handling
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
DOTInformation : DOT Class: 9; Packaging Group: III
EINECSNumber : 231-847-6
RTECSNumber : GL8800000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only