Please login to your online account to display your discounted pricing

Copper(II) selenite hydrate, 97%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Copper(II) selenite hydrate
CAS 10214-40-1
Physical Form Crystalline Powder
Assay Percent Range 97%
Molecular Formula CuO3Se·xH2O
Formula Weight 190.50 (Anhydrous)
MDL Number MFCD00010973
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4022106

  SDS
alfa aesar
4022106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4022118

  SDS
alfa aesar
4022118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4022130

  SDS
alfa aesar
4022130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.50 (anhydrous)
Quantity 5g
Color Blue
DOT Information DOT Class: 6.1, Packing Group: I
Physical Form Crystalline Powder
EINECS Number 233-526-6
CAS 10214-40-1
Assay Percent Range 97%
MDL Number MFCD00010973
Appearance Blue crystalline powder
Molecular Formula CuO3Se·xH2O
Health Hazard 1 H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
Chemical Name or Material Copper(II) selenite hydrate
Safety and Handling

H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.