missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) selenide, Black pieces, 99.5% (Metals basis), Alfa Aesar™

For Use in Semiconductors, As Catalyst in Kjeldahl Digestion

Manufacturer:  Alfa Aesar 1790122

 View more versions of this product

Catalog No. AA1790122



Description

Description

Form: 6mm and down pieces
Specifications

Specifications

Copper(II) selenide
99.5% (Metals basis)
MFCD00016058
Pieces
100g
1317-41-5
CuSe
142.5
Black

Chemical Identifiers

1317-41-5
CuSe
MFCD00016058
Safety and Handling

Safety and Handling

When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.

H301-H331-H373-H400-H410; Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-272-8

SDS Documents