missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) phthalocyanine, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04365018

 View more versions of this product

Catalog No. AA4365018



Description

Description

Copper(II) phthalocyanine are involved in the study of photosensitizer chemistry for uniform polymerization, luminescence chemistry and spectrophotometric analysis, organic synthesis and polymerization. This pigment is used in enamels, linoleum, inks, plastics, and rubber goods. Photoisomerizable phthalocyanines are used in rewritable CD or DVD printing. Other applications in organic solar cells, biosensitizers and display devices such as OLED , OTFT, Wearable Display, and e-paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper (II) phthalocyanine
350°C
C32H16CuN8
50 g
14,2520
It is insoluble in water.
Powder
147-14-8
Blue to Purple
MFCD00010719
4121848
C.I. 74160; Pigment Blue 15
576.08

Chemical Identifiers

147-14-8
C32H16CuN8
MFCD00010719
C.I. 74160; Pigment Blue 15
Safety and Handling

Safety and Handling

EINECSNumber : 205-685-1

RTECSNumber : GL8510000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only