Learn More
Copper(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01165130
Description
Copper(II) perchlorate hexahydrate is a strong oxidizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper(II) perchlorate hexahydrate | |
10294-46-9 | |
250 g | |
MFCD00149666 | |
14,2647 | |
Soluble in water, methanol, ethanol, acetic acid, acetic anhydride, and acetone | |
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2] | |
370.528 | |
370.53 (262.44 Anhydrous) | |
Odorless | |
Hygroscopic |
82°C | |
Crystalline | |
Cl2CuH12O14 | |
UN1481 | |
copper ii perchlorate hexahydrate, cupric perchlorate hexahydrate, unii-57jsh2lo1g, 57jsh2lo1g, copper diperchlorate hexahydrate, cupric diperchlorate hexahydrate, copper 2+ perchlorate hexahydrate, cupric perchlorate hexahydrate mi, acmc-20aldl, copper perchlorate cu clo4 2 hexahydrate | |
NHELIHXBJRANPL-UHFFFAOYSA-L | |
copper;diperchlorate;hexahydrate | |
12846356 | |
Reagent | |
1.505 | |
2.23 g/mL |
Chemical Identifiers
10294-46-9 | |
370.528 | |
NHELIHXBJRANPL-UHFFFAOYSA-L | |
12846356 | |
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2] |
Cl2CuH12O14 | |
MFCD00149666 | |
copper ii perchlorate hexahydrate, cupric perchlorate hexahydrate, unii-57jsh2lo1g, 57jsh2lo1g, copper diperchlorate hexahydrate, cupric diperchlorate hexahydrate, copper 2+ perchlorate hexahydrate, cupric perchlorate hexahydrate mi, acmc-20aldl, copper perchlorate cu clo4 2 hexahydrate | |
copper;diperchlorate;hexahydrate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber : 237-391-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only