missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) perchlorate hexahydrate, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4431918

 View more versions of this product

Catalog No. AA4431918



Specifications

Specifications

Copper (II) perchlorate hexahydrate
(metals basis)
MFCD00149666
14,2647
Soluble in water and ethanol. Very soluble in ethyl alcohol and acetone.
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2]
370.528
370.53 (262.44 Anhydrous)
99.999%
Odorless
Hygroscopic
50g
10294-46-9
Cl2CuH12O14
UN1481
57jsh2lo1g
NHELIHXBJRANPL-UHFFFAOYSA-L
copper;diperchlorate;hexahydrate
12846356
Crystalline
2.23 g/mL
1.505
82°C

Chemical Identifiers

10294-46-9
370.528
NHELIHXBJRANPL-UHFFFAOYSA-L
12846356
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2]
Cl2CuH12O14
MFCD00149666
57jsh2lo1g
copper;diperchlorate;hexahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

EINECSNumber : 237-391-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents