Please login to your online account to display your discounted pricing

Copper(II) Oxide Powder, ACS Grade, For Analysis, EMSURE™, EMD Millipore™

$1,045.34 - $3,494.09

Chemical Identifiers

Chemical Name or Material Copper(II) Oxide Powder
CAS 1317-38-0
Purity Grade ACS Grade
Molecular Formula CuO (Hill)
Formula Weight 79.55
UN Number UN 3077
Synonym Copper monoxide granulated
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

M1027660100

 
emd millipore corporation
1027660100
Each for $1,045.34

M1027660500

 
emd millipore corporation
1027660500
Each for $3,494.09
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 79.55
DOT Information DOT Class: 9, Packaging Group: III
CAS 1317-38-0
UN Number UN 3077
Packaging Plastic bottle
Synonym Copper monoxide granulated
Health Hazard 1 H302: Harmful if swallowed.
H410: Very toxic to aquatic life with long lasting effects.
Molecular Formula CuO (Hill)
Appearance Black solid
Odor Odorless
Chemical Name or Material Copper(II) Oxide Powder
EINECS Number 215-269-1
Physical Form Solid

For analysis

Safety and Handling

H302: Harmful if swallowed.
H410: Very toxic to aquatic life with long lasting effects.