Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) oxide, ACS

Chemical Identifiers

Chemical Name or Material Copper(II) oxide
CAS 1317-38-0
Purity Grade ACS Grade
Formula Weight 79.55
MDL Number MFCD00010979
UN Number UN3077
Merck Index 14,2646
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1315222

  SDS
alfa aesar™
1315222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1315236

  SDS
alfa aesar™
1315236
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 79.55
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Copper(II) oxide
TSCA TSCA
Odor Odorless
Purity Grade ACS Grade
Physical Form Wire, Random Pieces
Refractive Index 2.63
MDL Number MFCD00010979
CAS 1317-38-0
UN Number UN3077
Merck Index 14,2646
Color Brown to black
Melting Point 1326°C
Quantity 100g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Density 6.3g/mL
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number GL7900000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.