missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) oxide, ACS

Manufacturer:  Alfa Aesar™ 1315222

 View more versions of this product

Catalog No. AA1315222



Specifications

Specifications

Copper(II) oxide
CuO
UN3077
cupric oxide, copper ii oxide, copper oxide, copper monoxide, copper oxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
O=[Cu]
79.545
79.55
ACS
Odorless
Brown to Black
100g
1317-38-0
MFCD00010979
14,2646
QPLDLSVMHZLSFG-UHFFFAOYSA-N
oxocopper
14829
Wire, Random Pieces
6.3g/mL
2.63
1326°C

Chemical Identifiers

1317-38-0
79.545
QPLDLSVMHZLSFG-UHFFFAOYSA-N
14829
O=[Cu]
CuO
MFCD00010979
cupric oxide, copper ii oxide, copper oxide, copper monoxide, copper oxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
oxocopper
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : GL7900000

TSCA : TSCA

Documents