missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) oxide, 99.9995% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4435606

 View more versions of this product

Catalog No. AA4435606



Specifications

Specifications

Copper(II) oxide
99.9995% (Metal basis)
MFCD00010979
banacobru ol
QPLDLSVMHZLSFG-UHFFFAOYSA-N
oxocopper
14829
Powder
Odorless
Black
5g
1317-38-0
CuO
Black powder
Practically insoluble in water,and alcohol. Soluble in dilute acids,alkali cyanides,and ammonium carbonate solution.
O=[Cu]
79.545
79.55
6.315 g/cm3
2.63
1326°C

Chemical Identifiers

1317-38-0
79.545
QPLDLSVMHZLSFG-UHFFFAOYSA-N
14829
O=[Cu]
CuO
MFCD00010979
banacobru ol
oxocopper
Safety and Handling

Safety and Handling

H400-H410-H302: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.

DOTInformation : DOT Class: 9, Packing Group: III

EINECSNumber : 215-269-1

RTECSNumber : GL7900000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents