missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, 99.7% (metals basis)

CAS: 1317-38-0 | CuO | 79.55 g/mol

Supplier:  Thermo Scientific Chemicals 04169236

 View more versions of this product

Catalog No. AA4169236


Only null left
Explore available promotions

Description

Description

Copper(II) oxide is used as an adsorbent and absorbent, pigment, antifouling agent, wood preservative, optical glass polishing agent, welding flux for bronze materials, agricultural chemical and abrasive. It is used in the preparation of intermediates, oxidizing/reducing agents, surface treating agents and process regulators. It finds use in the manufacture of rayons, phosphors, and dry battery cells. It serves as a catalyst in certain chemical reactions of industrial importance. It is also employed in chemical sensors and superconducting materials. Hazardous materials like cyanides, dioxins and halogenated hydrocarbons can be disposed through oxidation with copper oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(II) oxide
1317-38-0
Powder
99.7% (Metal basis)
MFCD00010979
Practically insoluble in water and alcohol. Soluble in dilute acids,alkali cyanides,and ammonium carbonate solution.
[O--].[Cu++]
79.55
79.55
2.63
6.315 g/cm3
1326°C
Black
500 g
CuO
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
KKCXRELNMOYFLS-UHFFFAOYSA-N
copper(2+) oxidandiide
14829
Odorless
Black powder

Chemical Identifiers

1317-38-0
79.55
KKCXRELNMOYFLS-UHFFFAOYSA-N
14829
[O--].[Cu++]
CuO
MFCD00010979
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
copper(2+) oxidandiide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H400-H410-H302: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.

DOTInformation : DOT Class: 9, Packing Group: III

EINECSNumber : 215-269-1

RTECSNumber : GL7900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only