Learn More
Copper(II) oxide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01229936
Description
As a pigment, in sweeting petroleum gases, in galvanic electrodes, metallurgical flux, as an optical glass polishing agent, and as a catalyst in organic reactionsCopper(II) oxide is used as an adsorbent and absorbent, pigment, antifouling agent, wood preservative, optical glass polishing agent, welding flux for bronze materials, agricultural chemical and abrasive. It is used in the preparation of intermediates, oxidizing/reducing agents, surface treating agents and process regulators. It finds use in the manufacture of rayons, phosphors, and dry battery cells. It serves as a catalyst in certain chemical reactions of industrial importance. It is also employed in chemical sensors and superconducting materials. Hazardous materials like cyanides, dioxins and halogenated hydrocarbons can be disposed through oxidation with copper oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper(II) oxide | |
1317-38-0 | |
Powder | |
97% | |
MFCD00010979 | |
Insoluble in water and alcohol. Soluble in dil. acids,NH4Cl and KCNPractically insoluble in water,and alcohol; soluble in dilute acids,alkali cyanides,and ammonium carbonate solution. | |
[O--].[Cu++] | |
79.55 | |
79.55 | |
Black powder |
1326°C | |
Black | |
500 g | |
CuO | |
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide | |
KKCXRELNMOYFLS-UHFFFAOYSA-N | |
copper(2+) oxidandiide | |
14829 | |
2.63 | |
6.315 g/cm3 |
Chemical Identifiers
1317-38-0 | |
79.55 | |
KKCXRELNMOYFLS-UHFFFAOYSA-N | |
14829 | |
[O--].[Cu++] |
CuO | |
MFCD00010979 | |
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide | |
copper(2+) oxidandiide |
Safety and Handling
H400-H410-H302; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 215-269-1
RTECSNumber : GL7900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only