missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) oxide, 97%, ACROS Organics™

Manufacturer:  Acros Organics 197445000

 View more versions of this product

Catalog No. AC197445000



Specifications

Specifications

Copper(II) oxide
1317-38-0
CuO
MFCD00010979
cupric oxide, copper ii oxide, copper oxide, copper monoxide, copper oxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
O=[Cu]
79.545
79.54
97%
Solubility in water: insoluble. Other solubilities: soluble in dil.acids - slowly soluble in nh3, soluble in alkali cyanides and (nh4)2co3 solutions, practically insoluble in alcohol
1326.0°C
98%
97%
CuO
15, 2636
QPLDLSVMHZLSFG-UHFFFAOYSA-N
oxocopper
14829
Powder
Plastic bottle
Black
500g

Chemical Identifiers

1317-38-0
79.545
QPLDLSVMHZLSFG-UHFFFAOYSA-N
14829
O=[Cu]
CuO
MFCD00010979
cupric oxide, copper ii oxide, copper oxide, copper monoxide, copper oxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
oxocopper
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Very toxic to aquatic life.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 215-269-1

RTECSNumber : GL7900000

TSCA : TSCA

SDS Documents