missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) oxalate hemihydrate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1536518*

 View more versions of this product

Catalog No. AAA1536518



Description

Description

Applications
Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

Solubility
Soluble in NH{4}OH

Notes
Store in cool, dry, and well-ventilated area. Store away from oxidizing agents. Incompatible with acitve metals.
Specifications

Specifications

Copper(II) Oxalate Hemihydrate
50g
C4H6Cu2O9
UN3288
cupric oxalate, copper oxalate hemihydrate
LIZLXPYGKGKSBP-UHFFFAOYSA-N
copper;oxalic acid;hydrate
122130000
Powder
5893-66-3
98%
MFCD00150451
14,2645
Soluble in NH4OH
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Cu].[Cu]
325.175
160.57 (151.51 Anhydrous)
98%

Chemical Identifiers

5893-66-3
325.175
LIZLXPYGKGKSBP-UHFFFAOYSA-N
122130000
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Cu].[Cu]
C4H6Cu2O9
MFCD00150451
cupric oxalate, copper oxalate hemihydrate
copper;oxalic acid;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312

GHS P Statement
P280-P264-P270-P301+P312-P312-P363-P322-P302+P352-P330-P501a

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Documents