Please login to your online account to display your discounted pricing

Copper(II) nitrate trihydrate, 99%, for analysis, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  207681000

 View more versions of this product

Catalog No. AC207681000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay 98.5 to 104.0% (Iodometry)
Formula Weight 241.6
Physical Form Fine Powder
Quantity 100g
Color Cream to brown
Fieser 17,366
Merck Index 15, 2633
Purity Grade For analysis
RTECS Number GL7875000
Lead (Pb) 50ppm max.
Synonym Cupric nitrate
Chloride 20ppm max.
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment.
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. May intensify fire; oxidizer.
Health Hazard 1 Danger
MDL Number MFCD00149671
Chemical Name or Material Copper(II) nitrate trihydrate
Molecular Formula CuN2O6·3H2O
Assay Percent Range 99%
Name Note p.a.
Linear Formula Cu(NO3)2·3H2O
CAS 10031-43-3
Sulfate 50ppm max.
Chlorine (Cl) 20ppm max.
Packaging Plastic bottle
Iron (Fe) 50ppm max.
Trace Analysis 50ppm max.
Zinc (Zn) 50ppm max.
Calcium (Ca) 50ppm max.

Specifications

Chemical Name or Material Copper(II) nitrate trihydrate
Name Note p.a.
CAS 10031-43-3
Purity Grade For analysis
Assay 98.5 to 104.0% (Iodometry)
Assay Percent Range 99%
Molecular Formula CuN2O6·3H2O
Linear Formula Cu(NO3)2·3H2O
Formula Weight 241.6
MDL Number MFCD00149671
Synonym Cupric nitrate
Accessories

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment.

GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. May intensify fire; oxidizer.

SDS & Safety Documents