missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper (II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%, Alfa Aesar

For Use As a Catalyst, Nitrating Agent For Aromatic organosilicon Compounds, Nickel-Plating Baths

Manufacturer:  Alfa Aesar 1252336

 View more versions of this product

Catalog No. AA1252336



Specifications

Specifications

Copper(II) nitrate hemi(pentahydrate)
19004-19-4
CuN2O6·2.5H2O
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate, copper ii nitrate hemipentahydrate, copper ii nitrate hemipentahydrate
Crystalline Lump
2.32g/cm3
trihydrate
98.0-102.0%
MFCD00149671
232.59 (187.55 Anhydrous)
ACS
500g

Chemical Identifiers

19004-19-4
CuN2O6·2.5H2O
MFCD00149671
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate, copper ii nitrate hemipentahydrate, copper ii nitrate hemipentahydrate
Safety and Handling

Safety and Handling

H272-H314-H318-H302; May intensify fire; oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 221-838-5

storageNote1 : Hygroscopic

Documents