missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper (II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%, Alfa Aesar
For Use As a Catalyst, Nitrating Agent For Aromatic organosilicon Compounds, Nickel-Plating Baths
Manufacturer: Alfa Aesar 1252318
Specifications
Copper(II) nitrate hemi(pentahydrate) | |
19004-19-4 | |
CuN2O6·2.5H2O | |
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate, copper ii nitrate hemipentahydrate, copper ii nitrate hemipentahydrate | |
Crystalline Lump | |
2.32g/cm3 |
trihydrate | |
98.0-102.0% | |
MFCD00149671 | |
232.59 (187.55 Anhydrous) | |
ACS | |
50g |
Chemical Identifiers
19004-19-4 | |
CuN2O6·2.5H2O | |
MFCD00149671 | |
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate, copper ii nitrate hemipentahydrate, copper ii nitrate hemipentahydrate |
Safety and Handling
H272-H314-H318-H302; May intensify fire; oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.
DOTInformation : DOT Class: 5.1, Packaging Group: III
EINECSNumber : 221-838-5
storageNote1 : Hygroscopic