Please login to your online account to display your discounted pricing

Copper(II) nitrate, 2.5-hydrate, reagent ACS, +98%, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC405850025

 View more versions of this product

Catalog No. AC405850025

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Purity Grade ACS Reagent Grade
Formula Weight 232.6
Quantity 2.5kg
Color Blue
Assay 98.0 to 102.0%
Fieser 17,366
Merck Index 10, 2633
Nickel (Ni) 100ppm max.
Packaging Plastic bottle
Sodium (Na) 100ppm max.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May intensify fire; oxidizer.
Health Hazard 1 Danger
CAS 19004-19-4
Molecular Formula 2·5 H2O
MDL Number MFCD02253001
Assay Percent Range 98+%
Linear Formula 2·5H2O
Chemical Name or Material Copper(II) nitrate, hemipentahydrate
Potassium (K) 50ppm max.
Chlorine (Cl) 20ppm max.
Calcium (Ca) 50ppm max.
Synonym Cupric nitrate
Lead (Pb) 10ppm max.
Insoluble Matter 0.01% max.
Sulfate 100ppm max.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Iron (Fe) 50ppm max.

Specifications

Chemical Name or Material Copper(II) nitrate, hemipentahydrate
CAS 19004-19-4
Purity Grade ACS Reagent Grade
Assay 98.0 to 102.0%
Assay Percent Range 98+%
Molecular Formula 2·5 H2O
Linear Formula 2·5H2O
Formula Weight 232.6
MDL Number MFCD02253001
Synonym Cupric nitrate
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May intensify fire; oxidizer.

SDS & Safety Documents