missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) nitrate, 2.5-hydrate, reagent ACS, +98%, ACROS Organics™

Manufacturer:  Acros Organics AC405851000

 View more versions of this product

Catalog No. AC405851000



Specifications

Specifications

Copper(II) nitrate, hemipentahydrate
98+%
2·5H2O
17,366
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate, copper ii nitrate hemipentahydrate, copper ii nitrate hemipentahydrate
Crystalline Powder
ACS Reagent
20ppm max.
50ppm max.
100ppm max.
50ppm max.
Solubility in water: soluble.
Blue
100g
19004-19-4
2·5 H2O
MFCD02253001
10, 2633
232.6
≥98%
50ppm max.
0.01% max.
10ppm max.
Plastic bottle
100ppm max.
100ppm max.
114.0°C

Chemical Identifiers

19004-19-4
2·5 H2O
MFCD02253001
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate, copper ii nitrate hemipentahydrate, copper ii nitrate hemipentahydrate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents