Please login to your online account to display your discounted pricing

Copper(II) nitrate, 2.5-hydrate, reagent ACS, +98%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  405855000

 View more versions of this product

Catalog No. AC405855000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Purity Grade ACS Reagent Grade
Formula Weight 232.6
Quantity 500g
Color Blue
Fieser 17,366
Merck Index 10, 2633
Assay 98.0 to 102.0%
Nickel (Ni) 100ppm max.
Packaging Plastic bottle
Lead (Pb) 10ppm max.
Synonym Cupric nitrate
Chlorine (Cl) 20ppm max.
Potassium (K) 50ppm max.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May intensify fire; oxidizer.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger
Linear Formula 2·5H2O
MDL Number MFCD02253001
Chemical Name or Material Copper(II) nitrate, hemipentahydrate
CAS 19004-19-4
Assay Percent Range 98+%
Molecular Formula 2·5 H2O
Sodium (Na) 100ppm max.
Calcium (Ca) 50ppm max.
Sulfate 100ppm max.
Iron (Fe) 50ppm max.
Insoluble Matter 0.01% max.

Specifications

Chemical Name or Material Copper(II) nitrate, hemipentahydrate
CAS 19004-19-4
Purity Grade ACS Reagent Grade
Assay 98.0 to 102.0%
Assay Percent Range 98+%
Molecular Formula 2·5 H2O
Linear Formula 2·5H2O
Formula Weight 232.6
MDL Number MFCD02253001
Synonym Cupric nitrate
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May intensify fire; oxidizer.

SDS & Safety Documents