missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) methoxide, typically 98% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01886114

 View more versions of this product

Catalog No. AA1886114



Description

Description

Copper(II) methoxide is used as primary and secondary intermediates. Treatment of these methoxide derivatives with excess water results in the clean formation of mononuclear copper(II) hydroxide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(II) methoxide
∼206°C (decomposition)
Alcohol-like
Cu(OCH3)2
25 g
Moisture sensitive
125.61
Powder
1184-54-9
Green
(metals basis)
MFCD00015611
UN3181
Reacts with water.
98% (typically)

Chemical Identifiers

1184-54-9
MFCD00015611
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H228-H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only