missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper(II) hexafluoro-2,4-pentanedionate hydrate, Thermo Scientific Chemicals
$60.11 - $60.11
Chemical Identifiers
CAS | 155640-85-0 |
---|---|
Molecular Formula | C10H2CuF12O4 |
Molecular Weight (g/mol) | 477.65 |
MDL Number | MFCD00151019 |
InChI Key | HZXGNBMOOYOYIS-PAMPIZDHSA-L |
Synonym | copper ii hexafluoroacetylacetonate hydrate, bis hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoro-2,4-pentanedionate hydrate, copper hexafluoroacetylacetonate hydrate, copper ii hexafluoroacetyl-acetonate hydrate, hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoroacetylacetonate hydrate, elec. gr., bis 1,1,1,5,5,5-hexafluoroacetylacetonato copper ii hydrate, copper ii 1,1,1,5,5,5-hexafluoro-2,4-pentanedionate hydrate, 3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy cuprio oxy pent-3-en-2-one hydrate |
PubChem CID | 56845342 |
IUPAC Name | copper;(E)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate |
SMILES | [Cu++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1450503
|
Thermo Scientific Chemicals
01450503 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
155640-85-0 | |
477.65 | |
HZXGNBMOOYOYIS-PAMPIZDHSA-L | |
56845342 | |
[Cu++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F |
C10H2CuF12O4 | |
MFCD00151019 | |
copper ii hexafluoroacetylacetonate hydrate, bis hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoro-2,4-pentanedionate hydrate, copper hexafluoroacetylacetonate hydrate, copper ii hexafluoroacetyl-acetonate hydrate, hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoroacetylacetonate hydrate, elec. gr., bis 1,1,1,5,5,5-hexafluoroacetylacetonato copper ii hydrate, copper ii 1,1,1,5,5,5-hexafluoro-2,4-pentanedionate hydrate, 3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy cuprio oxy pent-3-en-2-one hydrate | |
copper;(E)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate |
Specifications
155640-85-0 | |
220°C (decomposition) | |
MFCD00151019 | |
Hygroscopic | |
Soluble in methanol, acetone, toluene | |
[Cu++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F | |
477.65 | |
477.64 (Anhydrous) | |
Copper(II) hexafluoro-2,4-pentanedionate hydrate |
∼100°C | |
C10H2CuF12O4 | |
1 g | |
copper ii hexafluoroacetylacetonate hydrate, bis hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoro-2,4-pentanedionate hydrate, copper hexafluoroacetylacetonate hydrate, copper ii hexafluoroacetyl-acetonate hydrate, hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoroacetylacetonate hydrate, elec. gr., bis 1,1,1,5,5,5-hexafluoroacetylacetonato copper ii hydrate, copper ii 1,1,1,5,5,5-hexafluoro-2,4-pentanedionate hydrate, 3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy cuprio oxy pent-3-en-2-one hydrate | |
HZXGNBMOOYOYIS-PAMPIZDHSA-L | |
copper;(E)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate | |
56845342 | |
Crystalline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-850-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only