missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) gluconate

Manufacturer:  Alfa Aesar™ 1532530

 View more versions of this product

Catalog No. AA1532530



Specifications

Specifications

Copper(II) gluconate
527-09-3
C12H22CuO14
14,2640
OCUCCJIRFHNWBP-IYEMJOQQSA-L
copper;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
131854750
453.85
155°C to 157°C
250g
MFCD00075297
copper gluconate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Cu+2]
453.84
CHEBI:31431
Powder

Chemical Identifiers

527-09-3
453.84
OCUCCJIRFHNWBP-IYEMJOQQSA-L
131854750
copper;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C12H22CuO14
MFCD00075297
copper gluconate
CHEBI:31431
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Cu+2]
Safety and Handling

Safety and Handling

RTECSNumber : LZ5058000

TSCA : TSCA

SDS Documents