Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) fluoride hydrate, Puratronic™, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Copper(II) fluoride hydrate
CAS 13454-88-1
Purity Grade Puratronic™
Formula Weight 101.54 (Anhydrous)
MDL Number MFCD00010977
UN Number UN3288
Merck Index 14,2638
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1300104

 
alfa aesar™
1300104
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1300109

 
alfa aesar™
1300109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1300118

 
alfa aesar™
1300118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.54 (Anhydrous)
Chemical Name or Material Copper(II) fluoride hydrate
TSCA TSCA
Odor Odorless
Purity Grade Puratronic™
Physical Form Crystalline
Assay 99.99%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00010977
CAS 13454-88-1
UN Number UN3288
Merck Index 14,2638
Color Blue
Melting Point >130°C (dec.)
Quantity 2g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 2.934g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.