Learn More
Copper(II) fluoride, anhydrous, 99.5%, Thermo Scientific Chemicals
Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent
Supplier: Thermo Scientific Chemicals 01148918
Description
Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent.Copper(II) fluoride is used in the preparation of fluorobenzene by reacting with benzene. It is also used in galvanic cells, high-temperature fluorinating agent and fluxing iron castings. Further, it is used as an efficient catalyst in Chan-Lam-Evans coupling reaction to synthesis the diaryl- ether compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper(II) fluoride | |
Anhydrous | |
7789-19-7 | |
50 g | |
UN3260 | |
Solubility in water at 20° ≈ 4;7g/100mL | |
99.5% | |
Hygroscopic |
950°C (decomposition) | |
White | |
Powder | |
MFCD00010977 | |
14,2638 | |
101.54 | |
Odorless | |
4.85 g/mL |
Chemical Identifiers
7789-19-7 | |
MFCD00010977 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 232-147-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only