missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II)-ethylenediamine complex, 1M solution in water, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC258550025

 View more versions of this product

Catalog No. AC258550025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Copper(II)-ethylenediamine complex
>110°C
1.1
1M Solution in Water
C4H18CuN4O2
MFCD00274628
Solubility in water: soluble
217.76
1.1000g/mL
Glass bottle
2.5L
14552-35-3,7732-18-5
Cu(H2NCH2CH2NH2)2(OH)2
Bis(ethylenediamine)copper(II)hydroxide
217.76

Chemical Identifiers

14552-35-3
217.76
Bis(ethylenediamine)copper(II)hydroxide
C4H18CuN4O2
MFCD00274628
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 238-597-7

TSCA : TSCA

SDS Documents