missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) ethoxide, 98%

Manufacturer:  Alfa Aesar™ 4631622

 View more versions of this product

Catalog No. AA4631622



Description

Description

  • Packaged under Argon
Specifications

Specifications

Copper (II) ethoxide
2850-65-9
Green
C4H10CuO2
MFCD00015650
copper ii ethoxide, copper diethoxide, ethanol, copper 2 salt, copper ethoxide, cupric ethoxide, copper ii ethoxide, acmc-20aldp, copper ii ethoxide, copper 2+ bis ethoxide, copper 2+ ion bis ethoxide
CC[O-].CC[O-].[Cu+2]
153.668
153.67
~120°C (decomposition)
100g
Air & Moisture Sensitive
Cu(OC2H5)2
UN3181
CRCKGIUJMFFISH-UHFFFAOYSA-N
copper;ethanolate
14419876
98%

Chemical Identifiers

2850-65-9
153.668
CRCKGIUJMFFISH-UHFFFAOYSA-N
14419876
CC[O-].CC[O-].[Cu+2]
C4H10CuO2
MFCD00015650
copper ii ethoxide, copper diethoxide, ethanol, copper 2 salt, copper ethoxide, cupric ethoxide, copper ii ethoxide, acmc-20aldp, copper ii ethoxide, copper 2+ bis ethoxide, copper 2+ ion bis ethoxide
copper;ethanolate
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

storageNote1 :

  • Packaged under Argon

SDS Documents