Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) ethoxide

Chemical Identifiers

Chemical Name or Material Copper(II) ethoxide
CAS 2850-65-9
Linear Formula Cu(OC2H5)2
Formula Weight 153.67
MDL Number MFCD00015650
UN Number UN3181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1886202

  SDS
alfa aesar™
1886202
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1886206

  SDS
alfa aesar™
1886206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1886214

  SDS
alfa aesar™
1886214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.67
Melting Point ≈120°C (dec.)
Chemical Name or Material Copper(II) ethoxide
Health Hazard 1 Warning
CAS 2850-65-9
Physical Form Powder
Linear Formula Cu(OC2H5)2
UN Number UN3181
Health Hazard 2 GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color Green
Quantity 0.5g
Health Hazard 3 GHS P Statement
P210-P280h-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Odor Alcohol-like
MDL Number MFCD00015650
Sensitivity Moisture Sensitive
Safety and Handling

Warning

GHS P Statement
P210-P280h-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.