missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) cyclohexanebutyrate, AAS, Cu 15.8%

Manufacturer:  Alfa Aesar™ 8726206

 View more versions of this product

Catalog No. AA8726206



Specifications

Specifications

Copper (II) cyclohexanebutyrate
2218-80-6
Cu 15.8%
C20H36CuO4
Cyclohexanebutyric acid copper(II) salt; 4-Cyclohexylbutyric acid copper(II) salt
C1CCC(CC1)CCCC(=O)O.C1CCC(CC1)CCCC(=O)O.[Cu]
404.05
402.01
126°C (decomposition)
5g
AAS Grade
MFCD00036399
ZGMZTUJLZVTXNR-UHFFFAOYSA-N
copper;4-cyclohexylbutanoic acid
131876184
Powder

Chemical Identifiers

2218-80-6
404.05
ZGMZTUJLZVTXNR-UHFFFAOYSA-N
131876184
C1CCC(CC1)CCCC(=O)O.C1CCC(CC1)CCCC(=O)O.[Cu]
C20H36CuO4
MFCD00036399
Cyclohexanebutyric acid copper(II) salt; 4-Cyclohexylbutyric acid copper(II) salt
copper;4-cyclohexylbutanoic acid
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents