Please login to your online account to display your discounted pricing

Copper(II) chloride, ultra dry, 99.995% (metals basis), Alfa Aesar™

Air Sensitive and Hygroscopic. >300°C partial decomposition to CuCl and Cl2

$77.59 - $77.59

Chemical Identifiers

Chemical Name or Material Copper(II) chloride
CAS 7447-39-4
Physical Form Powder
Assay Percent Range 99.995% (Metal basis)
Molecular Formula CuCl2
Formula Weight 134.45
MDL Number MFCD00010972
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3567306

 
alfa aesar
3567306
5g Each for $77.59

AA3567314

 
alfa aesar
3567314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3567322

 
alfa aesar
3567322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.45
Quantity 5g
Color Yellow-brown
DOT Information DOT Class: 8, Packing Group: III
Density 3.386g/cm3
Chemical Name or Material Copper(II) chloride
EINECS Number 231-210-2
Odor Odorless
Physical Form Powder
Assay Percent Range 99.995% (Metal basis)
Molecular Formula CuCl2
Appearance Yellow-brown powder
MDL Number MFCD00010972
CAS 7447-39-4
Health Hazard 1 H301-H314-H318-H400-H410: Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
Safety and Handling

H301-H314-H318-H400-H410: Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.