missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) chloride hydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1069822

 View more versions of this product

Catalog No. AA1069822



Specifications

Specifications

Copper(II) chloride hydrate
(metals basis)
MFCD00149674
14,2633
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
dichlorocopper;dihydrate
61482
134.45 (Anhydrous)
99.999%
2.510g/mL
Air Sensitive & Hygroscopic
100°C (decomposition)
100g
10125-13-0
Cl2CuH4O2
UN2802
caswell no. 235a
O.O.Cl[Cu]Cl
170.476
CHEBI:86318
Crystalline Aggregates
Puratronic™
Odorless
Blue-Green
3.0 to 3.8

Chemical Identifiers

10125-13-0
170.476
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
61482
dichlorocopper;dihydrate
Cl2CuH4O2
MFCD00149674
caswell no. 235a
CHEBI:86318
O.O.Cl[Cu]Cl
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: COPPER CHLORIDE

RTECSNumber : GL7030000

TSCA : TSCA

Documents