missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) chloride dihydrate, ACS, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 12458A3

 View more versions of this product

Catalog No. AA12458A3



Specifications

Specifications

Copper(II) chloride dihydrate
>99%
MFCD00149674
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
dichlorocopper;dihydrate
61482
170.48 (134.45 Anhydrous)
2.51g/mL
10125-13-0
Cl2CuH4O2
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, copper ii chloride dihydrate, caswell no. 235a, copper ii chloride dihydrate, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
O.O.Cl[Cu]Cl
170.476
CHEBI:86318
Granular
2kg

Chemical Identifiers

10125-13-0
170.476
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
61482
dichlorocopper;dihydrate
Cl2CuH4O2
MFCD00149674
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, copper ii chloride dihydrate, caswell no. 235a, copper ii chloride dihydrate, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
CHEBI:86318
O.O.Cl[Cu]Cl
Safety and Handling

Safety and Handling

EINECSNumber : 231-210-2

Documents