Learn More
Copper(II) chloride dihydrate, ACS, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01245822
Description
Copper(II) chloride is used as a catalyst for organic and inorganic reactions, mordant for dyeing and printing materials, pigment for glass, ceramics, as wood preservative, as disinfectant, and as a catalyst in the output of chlorine from hydrogen chloride. It is likewise employed in the oil industry as a purifying agent, in the manufacture of laundry marking inks, in metallurgy to recover mercury from ores, in refining copper, silver and gold, in tinting baths of iron and tin, in photography, in pyrotechnics, and to get rid of lead compounds from gasoline and oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper(II) chloride dihydrate | |
10125-13-0 | |
100 g | |
Cl2CuH4O2 | |
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701 | |
MPTQRFCYZCXJFQ-UHFFFAOYSA-L | |
copper(2+) dihydrate dichloride | |
61482 | |
170.48 (134.45Anhydrous) | |
Air sensitive; Hygroscopic |
100°C (decomposition) | |
Granules | |
>99% | |
MFCD00149674 | |
Soluble in water,methanol and ethanol. | |
O.O.[Cl-].[Cl-].[Cu++] | |
170.48 | |
CHEBI:86318 | |
ACS Reagent | |
2.51 g/mL |
Chemical Identifiers
10125-13-0 | |
170.48 | |
MPTQRFCYZCXJFQ-UHFFFAOYSA-L | |
61482 | |
copper(2+) dihydrate dichloride |
Cl2CuH4O2 | |
MFCD00149674 | |
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701 | |
CHEBI:86318 | |
O.O.[Cl-].[Cl-].[Cu++] |
Safety and Handling
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H318
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 231-210-2
RTECSNumber : GL7030000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only