missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper(II) chloride, anhydrous, 98% min., Alfa Aesar™
For Isomerization and Cracking Catalyst, Mordant in Dyeing and Printing Fabrics, in Electroplating Baths For Plating Cu On Al
Manufacturer: Alfa Aesar 1245730
Specifications
Copper(II) chloride | |
7447-39-4 | |
CuCl2 | |
Yellow-brown powder | |
Powder | |
Brown-Yellow |
Anhydrous | |
98% min. | |
MFCD00010972 | |
134.45 | |
Odorless | |
250g |
Chemical Identifiers
7447-39-4 | |
CuCl2 | |
MFCD00010972 |
Safety and Handling
H301-H314-H318-H400-H410; Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOTInformation : DOT Class: 8, Packaging Group: III
EINECSNumber : 231-210-2