Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) acrylate, typically 95%

$50.18 - $1,994.91

Chemical Identifiers

Chemical Name or Material Copper(II) acrylate
CAS 20074-76-4
Formula Weight 205.66
MDL Number MFCD00078371
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4177906

 
alfa aesar™
4177906
5g Each for $50.18

AA4177914

 
alfa aesar™
4177914
25g Each for $170.86

AA4177922

 
alfa aesar™
4177922
100g Each for $554.14

AA4177936

 
alfa aesar™
4177936
500g Each for $1,994.91
Description & Specifications

Specifications

Assay 95% (typically)
Formula Weight 205.66
Chemical Name or Material Copper(II) acrylate
Physical Form Powder
MDL Number MFCD00078371
CAS 20074-76-4
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Odor Weak
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.