missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) acetate monohydrate, ACS, 98.0-102.0%, Alfa Aesar

Manufacturer:  Alfa Aesar 1417722

 View more versions of this product

Catalog No. AA1417722



Specifications

Specifications

Copper(II) acetate monohydrate
6046-93-1
C4H8CuO5
Blue-green powder
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper;diacetate;hydrate
165397
Powder
1.882g/cm3
Blue-Green
monohydrate
98.0-102.0%
MFCD00149570
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
CC(=O)[O-].CC(=O)[O-].O.[Cu+2]
199.649
199.65 (181.64Anhydrous)
ACS
Odorless
100g

Chemical Identifiers

6046-93-1
199.649
NWFNSTOSIVLCJA-UHFFFAOYSA-L
165397
CC(=O)[O-].CC(=O)[O-].O.[Cu+2]
C4H8CuO5
MFCD00149570
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
copper;diacetate;hydrate
Safety and Handling

Safety and Handling

H400-H410-H302-H315-H319-H335; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 205-553-3

Documents