Learn More
Copper(II) acetate, 99%, pure, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 405370500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Copper(II) acetate | |
142-71-2 | |
Crystalline Powder | |
99% | |
(CH3CO2)2Cu | |
15,2614 | |
Solubility in water: soluble | |
CC(=O)[O-].CC(=O)[O-].[Cu+2] | |
181.64 | |
181.64 | |
Pure | |
Plastic bottle |
115.0°C | |
Blue to Green | |
50 g | |
C4H6CuO4 | |
01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 15,99 | |
copper ii acetate, cupric acetate, copper diacetate, copper acetate, venus copper, copper 2+ diacetate, cupric diacetate, neutral verdigris, crystals of venus | |
OPQARKPSCNTWTJ-UHFFFAOYSA-L | |
copper;diacetate | |
8895 | |
99% | |
Authentic |
Chemical Identifiers
142-71-2 | |
181.64 | |
copper ii acetate, cupric acetate, copper diacetate, copper acetate, venus copper, copper 2+ diacetate, cupric diacetate, neutral verdigris, crystals of venus | |
copper;diacetate |
C4H6CuO4 | |
OPQARKPSCNTWTJ-UHFFFAOYSA-L | |
8895 | |
CC(=O)[O-].CC(=O)[O-].[Cu+2] |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 205-553-3
RUO – Research Use Only