missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine, TCI America™

Manufacturer:  TCI America C30511G

 View more versions of this product

Catalog No. C30511G



Specifications

Specifications

Copper(II) 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine
1g
C80H88CuN8O8
5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine Copper(II)
CCCCOC1=C2C=CC=CC2=C(C3=C4[N-]C(=C31)N=C5C6=C(C7=CC=CC=C7C(=C6C(=NC8=NC(=NC9=NC(=N4)C1=C(C2=CC=CC=C2C(=C19)OCCCC)OCCCC)C1=C(C2=CC=CC=C2C(=C18)OCCCC)OCCCC)[N-]5)OCCCC)OCCCC)OCCCC.[Cu+2]
22834032
Crystal-Powder at 20°C
Undesignated
155773-67-4
MFCD00191950
JJIWQZDKAOMTKU-UHFFFAOYSA-N
1353.178
1353.18
≥97.0% (N,T)

Chemical Identifiers

155773-67-4
1353.178
JJIWQZDKAOMTKU-UHFFFAOYSA-N
22834032
C80H88CuN8O8
MFCD00191950
5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine Copper(II)
CCCCOC1=C2C=CC=CC2=C(C3=C4[N-]C(=C31)N=C5C6=C(C7=CC=CC=C7C(=C6C(=NC8=NC(=NC9=NC(=N4)C1=C(C2=CC=CC=C2C(=C19)OCCCC)OCCCC)C1=C(C2=CC=CC=C2C(=C18)OCCCC)OCCCC)[N-]5)OCCCC)OCCCC)OCCCC.[Cu+2]
Safety and Handling

Safety and Handling

TSCA : No

Documents