missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) 2-ethylhexanoate

Manufacturer:  Alfa Aesar™ 2053909

 View more versions of this product

Catalog No. AA2053909



Specifications

Specifications

Copper(II) 2-ethylhexanoate
149-11-1
Blue-Green
MFCD00015695
SZXCZUDYSKBGSX-UHFFFAOYSA-N
copper;2-ethylhexanoic acid
85309523
Powder/Lump
252°C (decomposition)
10g
C16H32CuO4
copper bis 2-ethylhexanoate
CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Cu]
351.974
349.96

Chemical Identifiers

149-11-1
351.974
SZXCZUDYSKBGSX-UHFFFAOYSA-N
85309523
CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Cu]
C16H32CuO4
MFCD00015695
copper bis 2-ethylhexanoate
copper;2-ethylhexanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents