missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(II) 2,4-pentanedionate hydrate

Manufacturer:  Alfa Aesar™ 1253536

 View more versions of this product

Catalog No. AA1253536



Specifications

Specifications

Copper(II) 2,4-pentanedionate hydrate
13395-16-9
Weak
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
Powder
230°C
500g
MFCD00000016
261.76 (Anhydrous)

Chemical Identifiers

13395-16-9
MFCD00000016
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents