missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(I) trifluoromethanesulfonate benzene complex, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 371090050

 View more versions of this product

Catalog No. AC371090050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Copper(I) trifluoromethanesulfonate benzene complex
7°C
90%
C8H6Cu2F6O6S2
MFCD00043300
12,144; 16,99
Solubility in water: .. Other solubilities: soluble in benzene,toluene
503.33
42152-46-5
Authentic
Glass bottle
(CF3SO3Cu)2C6H6
5g
Trifluoromethanesulfonic acid, copper(I) salt, benzene complex
503.33
90%

Chemical Identifiers

42152-46-5
503.33
Trifluoromethanesulfonic acid, copper(I) salt, benzene complex
C8H6Cu2F6O6S2
MFCD00043300
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protecti

GHS Signal Word: Warning

EINECSNumber : 255-686-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only