Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(I) trifluoromethanesulfonate benzene complex (2:1)

Chemical Identifiers

Chemical Name or Material Copper(I) trifluoromethanesulfonate benzene complex (2:1)
CAS 37234-97-2
Formula Weight 503.33
MDL Number MFCD00043300
UN Number UN3181
Synonym Copper(I) triflate; Bis[copper(I) trifluoromethanesulfonate]
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1698603

  SDS
alfa aesar™
1698603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1698606

  SDS
alfa aesar™
1698606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1698614

  SDS
alfa aesar™
1698614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 503.33
Sensitivity Air & Moisture Sensitive
Chemical Name or Material Copper(I) trifluoromethanesulfonate benzene complex (2:1)
Health Hazard 2 GHS H Statement
H372-H228
Causes damage to organs through prolonged or repeated exposure.
Flammable solid.
MDL Number MFCD00043300
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Synonym Copper(I) triflate; Bis[copper(I) trifluoromethanesulfonate]
UN Number UN3181
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P260-P280-P240-P314-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Powder
CAS 37234-97-2
Quantity 1g
Solubility Information Soluble in hot benzene
Safety and Handling

Danger

GHS P Statement
P210-P260-P280-P240-P314-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H372-H228
Causes damage to organs through prolonged or repeated exposure.
Flammable solid.