missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(I) sulfide, 99.5% (metals basis), Alfa Aesar

For Methods: Luminous Paints, Electrodes, Solar Cells

Manufacturer:  Alfa Aesar 1471830

 View more versions of this product

Catalog No. AA1471830



Specifications

Specifications

Copper(I) sulfide
99.5% (Metal basis)
MFCD00016062
copper i sulfide, cuprasulfide, copper sulfide cu2s, dicopper sulfide, dicopper sulphide, dicopper monosulfide, cuprous sulfide cu2s, unii-349m3c1rs1, cu2s, cupriosulfanyl copper
[SH-].[Cu].[Cu+]
160.16
159.14
Odorless
250g
22205-45-4
Cu2HS
Black powder
AQKDYYAZGHBAPR-UHFFFAOYSA-M
copper;copper(1+);sulfanide
62755
Powder
Black

Chemical Identifiers

22205-45-4
160.16
AQKDYYAZGHBAPR-UHFFFAOYSA-M
62755
[SH-].[Cu].[Cu+]
Cu2HS
MFCD00016062
copper i sulfide, cuprasulfide, copper sulfide cu2s, dicopper sulfide, dicopper sulphide, dicopper monosulfide, cuprous sulfide cu2s, unii-349m3c1rs1, cu2s, cupriosulfanyl copper
copper;copper(1+);sulfanide
Safety and Handling

Safety and Handling

EINECSNumber : 244-842-9

SDS Documents