missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(I) phenylacetylide

Manufacturer:  Alfa Aesar™ 17538ME

 View more versions of this product

Catalog No. AA17538ME



Specifications

Specifications

Copper(I) phenylacetylide
500mg
Air & Moisture Sensitive
MFCD00015578
copper 1+ ; ethynylbenzene, copper i phenylacetylide, copper i phenylacetylide, min
SFLVZUJLUPAJNE-UHFFFAOYSA-N
copper(1+);ethynylbenzene
6101206
Powder
13146-23-1
Yellow
C8H5Cu
UN3088
Soluble in acetone and acetonitrile; Insoluble in water
[C-]#CC1=CC=CC=C1.[Cu+]
164.674
153.68

Chemical Identifiers

13146-23-1
164.674
SFLVZUJLUPAJNE-UHFFFAOYSA-N
6101206
[C-]#CC1=CC=CC=C1.[Cu+]
C8H5Cu
MFCD00015578
copper 1+ ; ethynylbenzene, copper i phenylacetylide, copper i phenylacetylide, min
copper(1+);ethynylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H251-H260-H314-H318
Self-heating
may catch fire.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packaging Group: II; Proper Shipping Name: SELF-HEATING SOLID, ORGANIC, N.O.S.

SDS Documents