Please login to your online account to display your discounted pricing

Copper(I) oxide, 99% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Copper(I) oxide
CAS 1317-39-1
Molecular Formula Cu2O
Formula Weight 143.08
MDL Number MFCD00010974
UN Number UN3077
Merck Index 14,2664
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA4018822

  SDS
alfa aesar
4018822
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.08
Quantity 100g
Chemical Name or Material Copper(I) oxide
TSCA TSCA
Mesh Size -200 Mesh
Odor Odorless
Physical Form Powder
Refractive Index 2.705
Assay 99%
Assay Percent Notes (metals basis)
Molecular Formula Cu2O
MDL Number MFCD00010974
CAS 1317-39-1
Merck Index 14,2664
Boiling Point 1800°C (dec.)
Color Red to Dark Red
Melting Point 1235°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302;
Harmful if swallowed.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number GL8050000
Sensitivity Air and Moisture Sensitive
UN Number UN3077
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a;
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 6.0
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a;
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302;
Harmful if swallowed.