Learn More
Copper(I) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04018914
Description
It is used chiefly as a pigment in ceramics and in antifouling paints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1800°C (decomposition) | |
1235°C | |
1317-39-1 | |
25 g | |
Cu2O | |
Insoluble in water but soluble in acids. | |
Odorless | |
Air and moisture sensitive | |
6 g/cm3 |
Copper(I) oxide | |
Purple-Red | |
Powder | |
99% (Metal basis) | |
MFCD00010974 | |
143.08 | |
2.705 | |
Purple-red powder |
Chemical Identifiers
1317-39-1 | |
Cu2O | |
MFCD00010974 |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c
H302+H332-H318
DOTInformation : DOT Class: 9, Packing Group: III
EINECSNumber : 215-270-7
RTECSNumber : GL8050000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only